VWR®, Fern-Temperaturfühler für VWR Kühler
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
461-0101EA 411 EUR InStock
461-0101 461-0102 461-0103
VWR®, Fern-Temperaturfühler für VWR Kühler
Temperaturfühler
Remote temperature probes for circulator use with a chiller. Allows points of control to be switched from inside a chiller to a remote location. Compensates for heat loss through tubing between the circulator and the control point. Platinum RTD sensor with stainless steel sheath. Rubber coupling with 25,4 mm Ø, connects sheath to flexible cable. Probe cable ends in a DB9 female plug.
Länge×Ø: 152×4 mm
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge