General purpose UV cabinets, benchTop, UVC/T-M-AR
Lieferant: GRANT INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
UVC/T-M-ARUK UVC/T-M-AREURO UVCT-M-ARSKTUK
GRANUVC/T-M-ARUKEA 2900 EUR InStock
GRANUVC/T-M-ARUK 461-0822 GRANUVCT-M-ARSKTUK
General purpose UV cabinets, benchTop, UVC/T-M-AR
Dunstabzugshauben und Verkleidungen PCR-Arbeitsstationen und -Abzüge
Advanced benchtop UV cabinets providing aseptic conditions for a variety of biomedical and biochemical procedures. Innovative dual UV system: built-in UV-air recirculator provides constant decontamination of the air volume within the cabinet while working and traditional surface UV decontamination while the door is closed.
  • UV surface irradiation – via single 25 W 254 nm ozone free, open UV lamp
  • Patented built-in UV cleaner - recirculator with 25m³/hour continuous air flow, prevents unwanted contamination and protects the user from direct UV light during manipulation
  • Second UV lamp disinfects the working area, inactivating DNA/RNA fragments during 15 to 30 min of exposure; automatic switch-off when door is opened
  • Convenient, easy to use digital timer for accurate control of UV exposure
  • Built-in power socket
  • Quiet operation (33 to 37 dBa) and low energy consumption (67 W)
  • Front opening with three adjustable positions
The UV cleaner-recirculator consists of UV lamp, dust filters and a fan. The recirculation effect increases the maximum density of the UV light making it sufficiently effective for DNA / RNA inactivation and provide protection against contamination during a variety of DNA/RNA procedures, with dual UV lamp protection.
Stainless steel sides and work surface, UV protective film on glass panels.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge