PCR UV cabinet – DNA/RNA, economy model, UVT-B-AR
Lieferant: GRANT INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
UVT-B-ARUK UVT-B-AREURO
732-2842EA 2350 EUR InStock
732-2842 461-0824
PCR UV cabinet – DNA/RNA, economy model, UVT-B-AR
Dunstabzugshauben und Verkleidungen PCR-Arbeitsstationen und -Abzüge
Advanced benchtop UV cabinet providing aseptic conditions for a variety of biomedical and biochemical procedures. The economy bench model is dedicated to the protection against contamination during a variety of DNA/RNA procedures, with dual UV lamp protection.
  • UV surface irradiation - via single 25 W 254 nm open UV lamp
  • Patented built-in UV cleaner - recirculator with 25 m³/hour continuous air flow, prevents unwanted contamination and protects the user from direct UV light during manipulation
  • Second UV lamp disinfects the working area, inactivating DNA/RNA fragments during 15 to 30 min of exposure; automatic switch-off when door is opened
  • Convenient, easy to use digital timer for accurate control of UV exposure
  • Units have a built-in power socket
  • Quiet operation (33 to 37 dBa) and low energy consumption (67 W)
  • Front opening with three adjustable positions
Construction: Shock proof glass front, stainless steel sides, metal framework and stainless steel work surface, UV protective film on glass panels
Innovative dual UV system: built-in UV-air recirculator provides constant decontamination of the air volume within the cabinet while working and traditional surface UV decontamination while the door is closed.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge