Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Einhängethermostate, CORIO™ C, CD und CP

Lieferant: JULABO GmbH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

9011000 9012000 9013000
461-0887EA 865 EUR InStock
461-0887 461-0892 JULA9013000
Einhängethermostate, CORIO™ C, CD und CP
Umwälzthermostate Offene Umwälzsysteme Thermostatische Regler
The powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision, and a wide working temperature range. The CORIO™ C immersion thermostat is the basic model, it is ideal for standard internal applications, the CORIO™ CD/CP models are more advanced and can be equipped with an optional pump set for temperature control of external applications. The jet nozzle allows continuous adjustment of the pump stream in the bath. The bath attachment clamp enables either circulator to be mounted on virtually any bath (rectangular or round tanks).

  • Precise temperature control
  • For bath tanks with a max. filling volume of 30 (C units) or 50 litres (CD/CP units)
  • Bright, white, easy to read display
  • Very quiet and easy to operate
  • CD version has a USB interface and pump change-over between internal and external circulation via a lever located directly below the display
  • CP units have powerful and infinitely adjustable pressure pump, RS232 interface for online communication and early warning system for low liquid level

Max. flow rate / max. pressure:
C: 6 L/min, 0,1 bar
CD: 15 L/min, 0,35 bar
CP: 8 - 27 L/min, 0,1 to 0,7 bar

Zertifizierungen: C-Modelle entsprechen Klasse I (NFL), CD/CP-Modelle entsprechen Klasse III (FL) gemäß DIN 12876-1.

Lieferumfang: Supplied with attachment clamp for round or rectangular baths (which can also be transformed into a mount for laboratory stands in a few simple steps). A wide range of optional accessories, such racks, bath lids, tubing, adapters and the optional pump set are available.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge