VWR®, External PT100 Probes for
Circulating Baths
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
461-1373EA 490 EUR InStock
461-1373 461-1374 461-1376 461-1377
VWR®, External PT100 Probes for
Circulating Baths
Temperaturfühler
External PT100 probes is for monitoring the temperature of an external application.
For use with: Circulating baths, refrigerated circulating baths, heating circulating baths, stainless steel open bath systems, polycarbonate open bath systems, specialty products, calibration bath, 75 L refrigerated circulating bath.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge