46120.22
46120.36
46120.A3
46120.22EA
50.9
EUR
InStock
46120.22
46120.36
46120.A3
Magnesiumbromid Hexahydrat ≥98%
Magnesiumbromid Hexahydrat
Purity: 98+%
CAS No.: 13446-53-2
Molecular Formula: Br2Mg.6H2O
Formula Weight: 292.22 (184.13anhy)
Physical Form: Crystalline
Density: 2.00
Appearance: Colourless
Odour: Odourless
Melting Point: 172°
Solubility: Soluble in water, somewhat soluble in alcohol
MDL No.: MFCD00149780