Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Baths, Stirred, Digital, Clifton, NE4-D Series

Lieferant: NICKEL ELECTRO

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

NE4-8D NE4-14D NE4-22D NE4-28D NE4-38D
462-0048EA 2790 EUR InStock
462-0048 462-0043 462-0045 462-0046 462-0111
Baths, Stirred, Digital, Clifton, NE4-D Series
Bäder Wasserbäder, mit Umwälzung
General purpose stirred water baths offering good temperature stability and uniformity for consistent, repeatable results. Operation below ambient is possible using an optional refrigerated dip cooler.

  • Choice of five tank sizes and models with control from –20 to +99,9 °C with optional immersion cooler
  • PID temperature control with smart technology for adaptive or optimised temperature modes
  • Continuous or timer mode from 0 to 999 min with automatic switch-off and audible alarm
  • Low liquid level protection and auto setting temperature alarms
  • Anti-bacterial paint finish that actively inhibits bacterial growth

All tanks are manufactured from stainless steel and are totally crevice free for easy cleaning. The controller features a digital temperature display with a 0,1 °C resolution, and the powerful stirrer motor ensures good temperature stability throughout the tank.

Lieferumfang: Supplied with digital thermostirrer and removable stainless steel false base, but without racks or lids (lids are recommended when working above 60 °C or below ambient). Optional dip coller is required for operating at below ambient temperature. Please order required accessories separately.

Achtung! Bitte beachten: *RT = Ambient

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge