Baths, Thermostatically Controlled, Analogue, Clifton NE4-T Series
Lieferant: NICKEL ELECTRO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
NE4-14T NE4-22T NE4-28T NE4-8T
462-0108EA 2330 EUR InStock
462-0108 462-0109 462-0110 462-0107
Baths, Thermostatically Controlled, Analogue, Clifton NE4-T Series
Umwälzthermostate Offene Umwälzsysteme Bäder mit abnehmbarem Umwälzthermostat
These thermostatically controlled analogue stirred baths have a corrosion resistant stainless steel tank and durable outer case. Immersed components are corrosion resistant.
  • Thermostatic control, easy to use dial setting, ambient 5 to 99 °C
  • Powerful stirring within the tank for good water circulation and temperature stability
  • Controls with an illuminated on/off switch and 'heater on' indicator
  • User resettable safety cut-out float switch for low liquid level protection
  • Anti-bacterial paint finish that actively inhibits bacterial growth
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge