Boiling Baths, Analogue, Clifton, NE1B Series
Lieferant: NICKEL ELECTRO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
NE1B-14 NE1B-28
462-0116EA 2840 EUR InStock
462-0116 462-0117
Boiling Baths, Analogue, Clifton, NE1B Series
Bäder Heizbäder
Designed for applications that require continuous boiling. Baths have a corrosion resistant stainless steel working environment, with heater and safety cut out concealed underneath, all housed in a durable, chemical resistant, powder coated paint finished outer case.
  • Integral constant level device for connection to water supply combats water evaporation and maintains boiling
  • Include 'low' height removable stainless steel shelf to increase working volume
  • Drain outlet on 28 litre model for convenient bath emptying
  • Boiling can be regulated from vigorous to a gentle simmer
  • Anti-bacterial paint finish that actively inhibits bacterial growth
Bestellinformation: All models supplied with removable stainless steel perforated shelf, but without lids (lids are recommended when working above 60 °C). NE1B-28 is supplied with drain outlet for an accessory drain tap. Please order required accessories separately.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge