Desiccator Cabinets
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DWA301
467-0004EA 425 EUR InStock
467-0004
Desiccator Cabinets
Exsikkatoren Trockenkammern
Clear, high impact polystyrene cabinets have an airtight door with foam rubber gasket.
  • Cabinets can be stacked to save space
Lieferumfang: Supplied with sealed jar of silica gel.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge