Desiccator cabinets, horizontal, Dry-Keeper™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H420580001
468-4122EA 4010 EUR InStock
468-4122
Desiccator cabinets, horizontal, Dry-Keeper™, SP Bel-Art
Exsikkatoren Trockenkammern
This clear and durable PVC desiccator cabinet has sleek black ABS accents, a horizontal profile, and movable shelves for adjusting dry storage space configuration for moisture-sensitive materials.
  • Generous two cubic feet interior volume
  • Perforated shelves facilitate full circulation of air
  • Shelf rails can be repositioned on any one of 8 steps set at 28 mm intervals
  • Large in-door dial hygrometer for easy visualization of rH%
This cabinet has a gasketed door which opens down and closes with two latches for a tight seal. Interior opening measures 45×23 cm.
Bestellinformation: Additional shelves, rails and desiccant are available on request.
Lieferumfang: Supplied with desiccant tray.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge