Natriumhydroxid concentrate for dilution to 1 N aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Lieferant: AVANTOR PERFORMANCE MATERIALS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Gefahr
4689.EA 34.98 EUR InStock
4689.
Natriumhydroxid concentrate for dilution to 1 N aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sodium hydroxide
(1 equiv.=40.00 g NaOH)
Formel: NaOH
MW: 40 g/mol
Siedepunkt: ∼105 °C (1013 hPa)
Melting Pt: 323
Dichte: 1.327 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627
Order Now
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use
Normality (by titrimetry) Passes Test
(Makes 1N solution after dilution to 1000 mL)
Low in Carbonate
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge