Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ultra-low temperature freezers, Revco™ RDE

Lieferant: REVCO TECHNOLOGIES

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

RDE30086FV
471-1241EA 14093 EUR InStock
471-1241
Ultra-low temperature freezers, Revco™ RDE
Gefriergeräte Ultratiefkühlgeräte
Thermo Scientific™ Revco™ RDE Series ultra-low freezers feature four upright models, maximizing storage capacity from 30,000 up to 60,000 2 ml vials. New sustainability features include natural refrigerants and water-blown foam, compliant with many sustainability standards.

  • Easy-to-use set-up wizard, event log, temperature mapping and data downloads
  • Capacitive touch even when using latex gloves
  • Touch-screen datalogging and temperature mapping eliminates the need for a separate chart recorder
  • Built-in USB port for exporting data
  • Advanced security features: alarm settings, password-protected controller, padlock-compatible doors and optional access card entry
  • Ergonomic design: comfortable door handle, eye-level touch-screen and shelf-level placement that do not require floor-level loading of racks
  • Five inner doors minimize ambient temperature during sample retrieval
  • Low peak variation helps ensure sample protection throughout entire cabinet
  • Fast door opening recovery in as little as 14 minutes
  • 0,93 °C steady-state temperature variation over time
  • Long warm-up time from −80 to −50 °C in 5 hours

Limit environmental impact and lower HVAC costs:
Uses as low as 11,2 kWh/day, depending on size
Less heat emission lowers HVAC costs
Natural refrigerants compliant with EU F-Gas regulation: R170 and R290
Environmentally-friendly, water-blown foam insulation
Manufactured in an award-winning, zero-waste to landfill facility

Four capacities: 300, 400, 500 and 600 2-inch (5 cm) box

Temperature: −50...−86 °C.

Zubehörinformationen: Field installed options and accessories must be installed by a qualified professional.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

SERVICELEISTUNGEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge