Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

General purpose upright freezers, TSG series

Lieferant: HERAEUS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

TSG700FESBV TSG700FESCV TSG1500FESCV TSG400FESBV TSG400FESCV TSG1500FESBV
471-1331EA 4011 EUR InStock
471-1331 471-1333 471-1322 471-1326 471-1328 471-1320
General purpose upright freezers, TSG series
Gefriergeräte Gefriergeräte für das Labor und allgemeine Anwendungen
Designed for everyday laboratory needs the Thermo Scientific™ TSG series general purpose laboratory freezers are designed with your needs in mind. The TSG series range of freezers feature the performance, security, and quality required for your laboratory applications.

  • Freezers: −20 °C set point, settable range −25 to −10 °C
  • Wide range of sizes from solid door 400 to 1500 L double door
  • Whisper-quiet operation of <60 dBA
  • Adaptive defrost
  • White painted interior and exterior
  • Temperature, door ajar, and power failure (with battery backup) alarms
  • Low heat output
  • Green, natural refrigerants
  • Keyed lock

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used commonly in the laboratory.

3 year warranty.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge