Unter Argon in Ampullen verpackt
47178.06
47178.14
47178.06EA
218
EUR
InStock
47178.06
47178.14
Holmiumtrichlorid, ultra trocken ≥99,95% (Metall-Basis)
Holmiumtrichlorid
Purity: 99.95%
CAS No.: 10138-62-2
Molecular Formula: HoCl3
Formula Weight: 271.29
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.70
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 718°
MDL No.: MFCD00011051