47223.14
47223.22
47223.14EA
98.1
EUR
InStock
47223.14
47223.22
Hämatoxylin Hydrat, (max. 8% H₂O) ≥80% (Trockengewicht), Pulver
Hämatoxylin Hydrat
Purity: 80%
CAS No.: 517-28-2
Molecular Formula: C16H14O6.xH2O
Formula Weight: 302.29(anhy)
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Cream to brown
Melting Point: ca 200° dec.
Beilstein Reference: 91399
MDL No.: MFCD00078111