Chemical indicator strip, Comply™
Lieferant: 3M MEDICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M1250
481-0844EA 119 EUR InStock
481-0844
Chemical indicator strip, Comply™
Sterilisationsindikatoren
This steam chemical indicator offers a distinct off-white to dark brown/black color change when exposed to steam sterilization.
  • Strip is perforated so it can be used at full or half length depending on pack size
  • For use as an internal pack monitor
  • Can be used in all 250 to 273 °F steam sterilization cycles
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge