Medium to Large Capacity Priorclaves
Lieferant: Priorclave
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
EH100-SMART EH150-SMART EH200-SMART EH320-SMART EV100-SMART EV150-SMART EH200-BASE EV150-BASE EH100-BASE EH150-BASE EV100-BASE EH320-BASE
PRIOEH100-SMARTEA 0 EUR InStock
PRIOEH100-SMART PRIOEH150-SMART PRIOEH200-SMART PRIOEH320-SMART PRIOEV100-SMART PRIOEV150-SMART 481-0915 481-0918 481-0913 481-0914 481-0917 481-0916
Medium to Large Capacity Priorclaves
Sterilisatoren Autoklaven (Dampfsterilisatoren)
These versatile autoclaves are fully equipped for use in laboratories throughout the food, drink, dairy, health and pharmaceutical industries and offer a wide range of options and accessories to further improve their performance to suit individual load types. This flexibility is enhanced by the TACTROL™ microprocessor control system, specially developed for laboratory autoclaves, with progamming ranging from simple cycles to fully featured multi-program operation with logged records. All Priorclaves come complete with Biomaster antimicrobial coating, helping improved laboratory hygiene; it is effective against all bacteria and fungi including MRSA and therefore helps minimise the risk of cross contamination.
  • 500 mm chamber diameter to increase useable load capacity and reduce loading height on top loading models
  • Quickseal II single action closure
  • Fan assisted cooling
  • Timed free steaming for improved air removal
  • Media warming and delayed start allow the user to have media ready to pour at the start of the day
  • Maximum temperature: 138 °C (2,5 bar)
  • Mounted on lockable castors
Non-vacuum models:
Ideal for all standard traditional laboratory sterilisation procedures. Water fill is manual; during the cycle condensate leaves the vessel via a silicone tube that is connected to a heat resistant bottle. Ideal for use with media or liquid loads.
Vacuum models:
Automatic pulsed free steaming to improve temperature distribution and enhance steam penetration
Vacuum cooling to decrease cycle time
5 program memory
All vacuum models require mains water supply for pump operation.
Note: The installation of these models to a vented, sealed drain is strongly recommended and is a requirement of BS2646 when sterilising waste loads. Power supply required: 415 V, 15 A, 50 Hz, 10,5 kW, three phase with neutral and earth.
Lieferumfang: Top loading models supplied with two or three stainless steel baskets (depending on model). Front loading models with two stainless steel shelves.
Achtung! Bitte beachten: * These items are best ordered with the autoclave. Although they can be retrofitted, a site visit from a service engineer will be required with an associated extra charge.
** Vacuum models only. These options must be ordered with the autoclave and cannot be retrofitted.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge