Steam indicator tape, Comply™
Lieferant: 3M MEDICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M1322-12
489-0109EA 17.8 EUR InStock
489-0109
Steam indicator tape, Comply™
Sterilisationsindikatoren
Indicator tapes are Class 1 process indicators used for exposure monitoring. They provide assurance to the operator handling the processed items that the pack has been exposed to the sterilisation process without the need to open the pack or check load control records. Optional tape dispensers are available for easy dispensing of indicator tapes.
  • Chemical process indicator changes colour when exposed to the steam sterilisation process to provide assurance that the packs were processed without the need to open the pack
  • Versatile tape adheres to all types of wraps
  • Allows the user to write on tapes
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge