Spore ampoules
Lieferant: CROSSTEX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SA1-15-06 SA1-50-06 SA1-15-04 SA1-50-04 SA1-15-05 SA1-50-05 SA1-NC-10
489-0113EA 269 EUR InStock
489-0113 489-0114 NAMSSA1-15-04 NAMSSA1-50-04 NAMSSA1-15-05 NAMSSA1-50-05 NAMSSA1-NC-10
Spore ampoules
Sterilisationsindikatoren
These spore ampoules are ideal for use in validation and routine monitoring of liquid steam sterilization cycles. Spore ampoules are manufactured using hermetically sealed Type I borosilicate glass containing an exclusively formulated Tryptic
Soy Broth (TSB) modified with pH indicator, and Geobacillus stearothermophilus spores. No activation is required; simply expose Spore Ampoules to sterilization process and incubate.
  • Reduced incubation time of 48 hours
  • Clear visible results as growth is evident by turbidity and/or a color shift of the media from purple to yellow
  • Saves time by eliminating aseptic culturing required with traditional spore strips
  • Unique packaging insert keeps ampoules in upright position and allows for quick reference to the number of units remaining
Lieferumfang: Each shipment is accompanied by a certificate of analysis and instructions for use
Achtung! Bitte beachten: Storage under refrigerated conditions (2 to 8 °C) is required. Do not freeze spore ampoules.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge