452080
610053
610050
442080
6100535
495030
452060
24124
459503
400210
495150
495170
400210S
400110
453010
459504
495040
495190
495200
453070
400170
495160
26124
24241
21241
24242
495020
453020
400020
493000
453060
463100
470300
452500
402500
453280
463090
400030
400230
432080
452080S
452100
453030
455208
470370
492000
495010
400240
LIOF452080EA
37.5
EUR
InStock
LIOF452080
LIOF610053
LIOF610050
LIOF442080
LIOF6100535
495030.
452060.
24124.
459503.
400210.
495150.
495170.
400210S
400110.
453010.
459504.
495040.
495190.
495200.
453070.
400170.
495160.
26124.
24241.
21241-VWR
LIOF24242
495020.
453020.
400020.
493000.
453060.
463100.
470300.
452500.
402500.
453280.
LIOF463090
LIOF400030
LIOF400230
LIOF432080
LIOF452080S
LIOF452100
LIOF453030
LIOF455208
LIOF470370
LIOF492000
LIOF495010
LIOF400240
Fluids and Culture Media for Sterility Testing of Pharmaceutical Products
Medien für die Mikrobiologie
Fertigmedien für die Mikrobiologie
Culture media, diluting and rinsing sterile solutions for sterility testing of pharmaceutical products by membrane filtration.
- Store at 10 to 25 °C away from light
- Shelf life: 2 years
- Packaging, formulation and quality control suitable for media fill trial in pharmaceutical industries
- Some products are available with double or triple irradiated wrapping
Fluid A is a washing solution for rinsing membranes during sterility testing.
Fluid D is a washing solution recommended for dissolving oils or materials containing lecithin or a preservative and can be used for devices labeled as 'sterile pathway'.
Fluid K is a sterile diluent used for rinsing membranes during sterility and is recommended for use with substances containing petrolatum/oil.
Fluid thioglycollate medium and Tryptic soy broth are general purpose liquid enrichment culture media used for sterility control of pharmaceutical products and for cultivation and isolation of fastidious anaerobic and aerobic microorganisms.
TSB is also available as triple-wrapped and gamma-irradiated bottles, particularly suitable for use in restricted areas and for aseptic process simulations (media fill trial) in the pharmaceutical industry.
Zertifizierungen: The composition is in accordance with the requirements of the Harmonised method in the United States (USP), European (EP) and Japanese (JP) Pharmacopoeias. Thioglycollate Medium is also in compliance with ISO 7937 for isolation of Clostridium perfringens.