Multi-element standard solution 3 for ICP, TraceCERT®, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
49596-100ML
49596-100MLEA 126 EUR InStock
49596-100ML
Multi-element standard solution 3 for ICP, TraceCERT®, Supelco®
Standards Kationischer Standards Mehrelement-Standards
Ca, 2000 mg/l
K, 200 mg/l
Mg, 400 mg/l
Na, 1000 mg/l
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge