Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

all-rac-α-Tocopherol 96,0-102,0%, EMPROVE® ESSENTIAL USP, Ph. Eur., BP, E307 suitable for use as excipient, SAFC®

Lieferant: MERCK PRODUCTION CHEMICALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

5.00949.5000 5.00949.9020
5.00949.5000EA 2300 EUR InStock
5.00949.5000 5.00949.9020
all-rac-α-Tocopherol 96,0-102,0%, EMPROVE® ESSENTIAL USP, Ph. Eur., BP, E307 suitable for use as excipient, SAFC®
DL-α-Tocopherol
Corresponds to Ph. Eur., BP, USP, E 307
Conforms to the purity criteria on food additives according to the current European Commission Regulation.
# specified acc. to EMEA/CHMP/SWP/4446/2000 (Specification limits for residues of metal catalysts or metal reagents).
Other residues of metal catalysts or metal reagents acc. to this guideline are not likely to be present.


Formel: C₂₉H₅₀O₂
MW: 430.71 g/mol
Storage Temperature: Raumtemperatur
MDL Number: MFCD00072051
CAS-Nummer: 10191-41-0
EINECS: 233-466-0
Order Now
Spezifikation Testergebnisse
Assay (GC, Ph. Eur.) 96.0 - 102.0 %
Assay (GC, USP) 96.0 - 102.0 %
Identity (IR) conforms
Identity (GC) conforms
Identity (colour reaction) conforms
Appearance clear, colourless to yellow-brownish, viscous, oily liquid; almost odourless
Acidity conforms
Optical rotation α 20/D (100 g/l; anhydrous ethanol) -0.01 - 0.01 °
Optical rotation α 25/D (100 g/l; chloroforme) -0.05 - 0.05 °
Refractive index (n 20/D) 1.503 - 1.507
Spec. Absorptivity A 1%/1cm (292 nm; 0,1 g/l, anhydrous ethanol) 72 - 76
Spec. Absorptivity A1%/1cm (292 nm; 0,05 g/l , anhydrous Ethanol) 71 - 76
Heavy metals (as Pb) ≤ 10 ppm
As (Arsenic) ≤ 3 ppm
Cu (Copper) ≤ 10 ppm
Hg (Mercury) ≤ 1 ppm
Pb (Lead) ≤ 2 ppm
Zn (Zinc)# ≤ 25 ppm
Heptane (HS-GC) ≤ 5000 ppm
Methanol (HS-GC) ≤ 3000 ppm
Pyridine (HS-GC) ≤ 200 ppm
Toluene (HS-GC) ≤ 890 ppm
Other residual solvents (ICH Q3C) excluded by manucfacturing process
Related substances (Ph. Eur., impurity A) ≤ 0.5 %
Related substances (Ph. Eur., impurity B) ≤ 1.5 %
Related substances (Ph. Eur., sum of impurities C and D) ≤ 1.0 %
Related substances (Ph. Eur., any other impurity) ≤ 0.25 %
Related substances (Ph. Eur., total impurities) ≤ 2.5 %
Sulfated ash (600 °C) ≤ 0.1 %

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge