DMNP-EDTA tetrapotassium salt (Caged Calcium) ≥90% (durch HPLC)
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
50050
50050.EA 621 EUR InStock
50050.
DMNP-EDTA tetrapotassium salt (Caged Calcium) ≥90% (durch HPLC)
DMNP-EDTA tetrapotassium salt (Caged Calcium)
DMNP-EDTA (also known as DM-Nitrophen™) is a caged Ca2+ chelator. Upon photolysis, the Kd for Ca2+ increases from 5 nM to 3 mM, resulting in a pulse of free Ca2+.
  • Light yellow solid soluble in water (pH >6) and DMSO
  • Store desiccated at -20°C and protect from light
  • Aqueous solutions stable for at least three months when stored at 4°C, protected from light
DM-Nitrophen is a trademark of MilliporeSigma.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge