Supelco® Ascentis® Express ES-Cyano HPLC and UHPLC Columns
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
53468-U 53470-U 53472-U 53473-U 53475-U 53476-U 53478-U 53479-U 53481-U 53483-U 53484-U 53486-U 53489-U 53491-U 53492-U 53494-U 53495-U 53496-U 53497-U 50557-U 50558-U 50559-U 50562-U 50563-U 50564-U 50566-U 50567-U 50568-U 50569-U 50570-U 50574-U 50575-U 50577-U 50581-U 50583-U 50585-U 50588-U 50591-U 50592-U 50593-U 50597-U 51709-U 51712-U 51717-U 51721-U 51724-U 51725-U 51727-U 51728-U 51729-U 51732-U 51734-U 51736-U 51739-U
53468-UEA 484 EUR InStock
53468-U 53470-U 53472-U 53473-U 53475-U 53476-U 53478-U 53479-U 53481-U 53483-U 53484-U 53486-U 53489-U 53491-U 53492-U 53494-U 53495-U 53496-U 53497-U 50557-U 50558-U 50559-U 50562-U 50563-U 50564-U 50566-U 50567-U 50568-U 50569-U 50570-U 50574-U 50575-U 50577-U 50581-U 50583-U 50585-U 50588-U 50591-U 50592-U 50593-U 50597-U 51709-U 51712-U 51717-U 51721-U 51724-U 51725-U 51727-U 51728-U 51729-U 51732-U 51734-U 51736-U 51739-U
Supelco® Ascentis® Express ES-Cyano HPLC and UHPLC Columns
Chromatographiesäulen
Ascentis® Express ES-Cyano high-speed, high-performance liquid chromatography columns are based on the highly efficient Fused-Core® particle design.
The Fused-core particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0,5 µm thick porous shell.
The sterically protected, extensively endcapped diisopropyl-cyanopropylsilane stationary phase of Ascentis Express ES-Cyano provides a stable, reversed-phase packing that can be used for basic, acidic or neutral compounds.
Lieferumfang: Supplied with column test certificate inside the box.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge