Organic reference standard, Bisphenol B, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
50877-100MG
50877-100MGEA 77.6 EUR InStock
50877-100MG
Organic reference standard, Bisphenol B, analytical standard, Supelco®
Standards Organische Standards
Bisphenol B is a congener of bisphenol A, used primarily for the manufacturing of phenolic resins and is also a potent food contaminant.
Bisphenol B may be used as a reference standard for the determination of bisphenol B in liquid food matrices and powdered infant formulae and human urine samples by using dispersive liquid-liquid microextraction (DLLME) with in situ derivatisation coupled with heart-cutting gas chromatography-mass spectrometry (GC-MS).
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5b3db7a65796aa3e11cbee95d533ca66097eb1e&vid=b5b3db7a65796aa3e11cbee95d533ca66097eb1e&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge