Phytochemical standard, Procyanidin C1, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
50878-5MG
50878-5MGEA 809 EUR InStock
50878-5MG
Phytochemical standard, Procyanidin C1, analytical standard, Supelco®
Standards Organische Standards
Procyanidins are classified under the class of flavan-3-ols which are the common components of plant-derived food products.
Procyanidin C1 can be used as an analytical reference standard for the determination of procyanidin C1 in cocoa-based products by ultra-high performance liquid chromatography coupled to quadrupole Orbitrap high resolution mass spectrometry (UHPLC-Q-Orbitrap-MS/MS), wild apple by UHPLC equipped with diode array detection (DAD) and high resolution multistage mass spectrometry (HRMS), food materials and biological samples by HPLC combined with fluorescence detector (FLD) as well as DADL, lentil seed coats by HPLC-MS with ultraviolet (UV) detection, grape seeds by high-speed counter-current chromatography (HSCCC).
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f97d3ce31e3f7d5b9f5f12a30de0fb0898348de&vid=8f97d3ce31e3f7d5b9f5f12a30de0fb0898348de&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge