VWR®, Analysensiebe, 100×45 mm
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
510-0526EA 194 EUR InStock
510-0526 510-0527 510-0528 510-0529 510-0530 510-0531 510-0532 510-0533 510-0534 510-0535 510-0536 510-0537 510-0500 510-0501 510-0502 510-0503 510-0504 510-0505 510-0506 510-0507 510-0508 510-0509 510-0510 510-0511 510-0512 510-0513 510-0514 510-0515 510-0516 510-0517 510-0518 510-0519 510-0520 510-0521 510-0522 510-0523 510-0524 510-0525 510-0538 510-0539 510-0540 510-0541 510-0542 510-0543 510-0544 510-0545 510-0546 510-0547 510-0548 510-0549 510-0550 510-0551 510-0552 510-0553
VWR®, Analysensiebe, 100×45 mm
Siebe Analysensiebe
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Lieferumfang: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
SERVICELEISTUNGEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge