VWR®, Analysensiebe, 200×32 mm
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
510-0608EA 984 EUR InStock
510-0608 510-0609 510-0610 510-0611 510-0612 510-0613 510-0614 510-0615 510-0616 510-0617 510-0618 510-0619 510-0620 510-0621 510-0622 510-0623 510-0624 510-0625 510-0626 510-0627 510-0628 510-0629 510-0630 510-0631 510-0632 510-0633 510-0634 510-0635 510-0636 510-0637 510-0638 510-0639 510-0640 510-0641 510-0642 510-0643 510-0644 510-0645 510-0646 510-0647 510-0648 510-0649 510-0650 510-0651 510-0652 510-0653 510-0654 510-0655 510-0656 510-0657 510-0658 510-0659 510-0660 510-0661 510-0662 510-0663 510-0664 510-0665 510-0666 510-0667 510-0668 510-0669 510-0670 510-0671 510-0672 510-0673 510-0674 510-0675 510-0676 510-0677 510-0678
VWR®, Analysensiebe, 200×32 mm
Siebe Analysensiebe
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Lieferumfang: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
SERVICELEISTUNGEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge