VWR®, Analysensiebe, 50×25 mm
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
510-1792EA 128 EUR InStock
510-1792 510-1828 510-1829 510-1793 510-1794 510-1795 510-1796 510-1797 510-1798 510-1799 510-1800 510-1801 510-1802 510-1803 510-1804 510-1805 510-1806 510-1807 510-1808 510-1809 510-1810 510-1811 510-1812 510-1813 510-1814 510-1815 510-1816 510-1817 510-1818 510-1819 510-1820 510-1821 510-1822 510-1823 510-1824 510-1825 510-1826 510-1827 510-1830 510-1831 510-1832 510-1833 510-1834 510-1835 510-1836 510-1837 510-1838 510-1839 510-1840 510-1841 510-1842 510-1843 510-1844 510-1845 510-1846 510-1847
VWR®, Analysensiebe, 50×25 mm
Siebe Analysensiebe
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3,1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request. Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2,1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Lieferumfang: Supplied as standard with a certificate of compliance 2,1 according to DIN EN 10204.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge