Vibratory sieve shakers, Analysette®3 spartan
Lieferant: FRITSCH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
03.8020.00
510-4102EA 2494 EUR InStock
510-4102
Vibratory sieve shakers, Analysette®3 spartan
Schüttler Siebmaschinen
Analysette®3 spartanis a vibratory sieve shaker for all typical sieving tasks in the laboratory, with optical adjustment of the amplitude on the running instrument. The Analysette®3 spartan is also suitable for quantitative particle size analysis of solids and suspensions of all kinds through dry or wet sieving with woven test sieves. It is convertible into a micro mill for fine comminution of dry laboratory samples or solids in suspensions and for homogenisation of emulsions or pastes.
  • Automatic sieve analysis with sieve evaluation software autosieve
  • Quick conversion to vibratory micro mill pulverisette
  • Exact entry of the sieving time via a precise digital timer
  • Manual adjustment of the amplitude during sieving
  • Display of the amplitude during sieving
  • Ergonomically installed robust soft touchpad
  • Optimum power consumption prevents warming up of the sieving system
Up to 10 test sieves can be used simultaneously per working cycle and this system also allows up to 5 sieving operations to be performed at the same time.
Bestellinformation: Supplied with Easytwist sieve stack tensioner with steel-reinforced plastic bands and autosieve software. Clamping lid, test sieves or sieve pan and other accessories need to be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge