Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

5(6)-Carboxyfluorescein ≥95% (durch HPLC) fluoreszierender pH-Indikator

Lieferant: BIOTIUM

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


51013
51013.EA 95.3 EUR InStock
51013.
5(6)-Carboxyfluorescein ≥95% (durch HPLC) fluoreszierender pH-Indikator
5(6)-Carboxyfluorescein
5-(and-6)-FAM (full name: 5-(and-6)-Carboxyfluorescein mixed isomer) has a pKa of 6.5 and can be used as a pH indicator. Its excitation spectrum and fluorescence response to pH are similar to those of BCECF. Carboxyfluorescein can also be used as a tracer dye. The dye is membrane-impermeant and can be loaded into cells by microinjection or scrape loading.

  • λExλEm (pH 9) = 492/514 nm
  • ε (pH 9) = 82,000
  • Cell permeability: Membrane impermeant
  • Cellular localization: Fluid phase tracer
  • pKa 6.5
  • Colors: Green
  • Yellow orange solid soluble in pH>6 water
  • Store at 4°C and protect from light, especially in solution


Formel: C₄₂H₂₄O₁₄
MDL Number: MFCD00151081
CAS-Nummer: 72088-94-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05754d8ccebfb8b75f45fa05fa1a5ade7b621942&vid=05754d8ccebfb8b75f45fa05fa1a5ade7b621942&prid=web&clid=SAPDAM
Order Now

 

PRODUKT

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge