Zentrifugaleinheiten, Vivaspin® 15R
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
VS15RH22 VS15RH21 VS15RH12 VS15RH11 VS15RH02 VS15RH01 VS15RH91 VS15RH92
512-4124EA 596 EUR InStock
512-4124 512-4123 512-4120 512-4119 512-4122 512-4121 512-4125 512-4126
Zentrifugaleinheiten, Vivaspin® 15R
Filter Zentrifugaleinheiten
Vivaspin® 15R is targeting the volume segment of 4 to 15 ml with the Hydrosart® regenerated cellulose membrane. This membrane is ideal when extremely high recovery rate and very low adsorption is required, for example in applications such as concentration and desalting of immunoglobulin fractions.
  • Highest recovery at low absorption (95 to 98%)
  • Extremely short concentration time (30-fold in 15 min.)
  • Very small holdup volume (<20 µl)
  • Crystal clear housing and volume graduations
  • Vertical twin membrane for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
Vivaspin® 15R has all the unique features of the Sartorius centrifugal concentrators, including a patented vertical twin membrane and a dead stop.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge