Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Glass microfibre filters, 934-AH™ RTU, Whatman™

Lieferant: GE Healthcare - Whatman

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

9907-042 9907-055 9907-047 9907-090 9907-9436 2827-047 3827-035 3827-042 3827-047 3827-070 3827-090 4827-035 4827-042 4827-047 4827-070 4827-090 9907-070 9927-047 9927-070 9927-090
513-0114EA 123 EUR InStock
513-0114 513-0116 513-0115 513-0117 WHAT9907-9436 513-0140 513-0127 513-0128 513-0129 513-0130 513-0131 513-0132 513-0133 513-0134 513-0135 513-0136 WHAT9907-070 513-0137 513-0138 513-0139
Glass microfibre filters, 934-AH™ RTU, Whatman™
Filter Glasfaserfilter
These 934-AH™ RTU grade filter papers have been pre-washed, dried (or ignited), cooled, desiccated and weighed, which eliminates the pre-treating steps that are otherwise required by the US EPA Laboratory Standard Method 2540 parts C, D, and E. These filter papers are widely used for dissolved, suspended and volatiles solid analysis. They are also used as a wastewater filter for a wide range of water monitoring applications including the monitoring of rivers, lakes and coastal waters, as well as analysing purified discharge of wastewater treatment plants and monitoring discharge water from industrial plants.

  • Superior fine particle retention at high flow rates
  • Capable of processing very turbid samples
  • Particle retention: 1,5 µm

934-AH™ RTU for suspended and dissolved solids: Pre-washed, dried, cooled, and weighed filters with certified mass loss between washing and drying steps of the lesser of less than 0,5 mg or 4% in accordance with US EPA Lab Standard Method 2540 parts C and D. Most filters come weighed to four decimal places, however a version of the 47 mm filter can be measured to five decimal places if needed.

934-AH™ RTU for volatiles: Pre-washed, pre-weighed and pre-fired at 550 °C aligned to US EPA Laboratory Standard Method 2540 parts C, D, and E.

934-AH™ RTU double weigh: Drying, cooling and weighing steps repeated twice for customers with the highest regulatory concern.

* Comes pre-washed and pre-weighed to 5 decimal places.

Verpackung: The pre-weighed filter papers are packaged within an aluminium pan with the filter weight clearly printed on a heat resistant label. Each pan has a unique barcode. Each box also has a barcode that can be used to download the weights of all filters from that box.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge