Microbiological filtration system, MBS II, Whatman™
Lieferant: GE Healthcare - Whatman
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10445901
514-0054EA 193 EUR InStock
514-0054
Microbiological filtration system, MBS II, Whatman™
Filter Membranfiltergeräte
The microbiological filtration system is used for optimal microbiological control using membranes. The design of the system, which consists of an electrical membrane dispenser, a funnel dispenser, and a vacuum manifold, leads to more reproducible results.
  • Shorter preparation time
  • High reproducibility
  • Large funnel capacity for foaming liquids
  • Funnels can be autoclaved up to 50 times
The special sealing technique ensures easy handling and a good integrity of the funnel and membrane during filtration. This reduces any cross contamination to a minimum.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge