Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Spritzenlose Filter, Mini-UniPrep™ G2, Whatman™

Lieferant: GE Healthcare - Whatman

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

GN203APEAQUSP GN203NPEAQU GN203NPEAQUSP GN203NPENYLSP GN203NPEORG GN203NPEPP GN203NPEPPSP GN203NPUAQU GN203NPUAQUSP GS203NPUORGSP GS203NPUGMFSP GN503NPUORG GS203NPUAQUSP GN503NPURC GS203NPEPPSP GS203NPEPP GS203NPEORG GS203NPENYLSP GS203NPENYL GS203NPEAQUSP GN203NPURC GN503NPUGMF GN503NPUAQU GN503NPERC GN503NPEPP GN503NPEORG GN503NPENYL GN203APEAQU GN203NPURCSP GS203NPEAQU GN203NPUGMF GN203NPERCSP GN203NPERC GN203NPENYL GN503NPEAQU GS203NPUAQU GS503NPUAQU GS503NPENYL GS503NPEAQU GS203NPUORG GS503NPUORG GS203NPUGMF
514-0385EA 559 EUR InStock
514-0385 514-0387 514-0388 514-0389 514-0390 514-0392 514-0393 514-0394 514-0395 514-0399 516-0476 514-8149 516-0474 514-8150 516-0472 516-0471 516-0470 516-0469 516-0468 516-0467 514-7999 514-8148 514-8147 514-8146 514-8145 514-8144 514-8143 514-8136 514-8141 516-0466 514-8139 514-8138 514-7998 514-7997 514-8142 516-0473 516-0481 516-0479 516-0478 516-0477 516-0482 516-0475
Spritzenlose Filter, Mini-UniPrep™ G2, Whatman™
Filter Spritzenlose Filter
Mini-UniPrep™ G2 is an 'all in one' syringeless filter with a borosilicate glass chamber. It replaces syringe filters for autosampler-supported UHPLC/HPLC sample preparation with easier handling and fewer consumable components.

  • Features an integrated autosampler vial, plunger with attached filter membrane, and septum/cap
  • Borosilicate glass vials eliminate plastic-based leachables that can originate from PP vials
  • Designed to hold up to 0,5 ml of unfiltered sample that can then be loaded directly into an autosampler
  • Easy to see when a sample has been filtered
  • Wide range of membranes with 0,2 and 0,45 µm pore sizes to meet specific requirements
  • Also available in amber glass for light-sensitive samples

A variety of filter membrane media are available to meet your needs. Browse the Whatman line to find the right filter and compressor for your application.

Mini-UniPrep™ syringeless filters combine a membrane filter, autosampler vial, septum, and plunger into a single device to provide efficient sample filtration for Ultra High Performance Liquid Chromatography (UHPLC)/High Performance Liquid Chromatography (HPLC) analysis.

Once compressed, Mini-UniPrep™ G2 can be placed into any autosampler that accepts a 12×32 mm autosampler vial for injection into your instrument. Alternatively, the septum can be pierced with a needle and the sample drawn off for manual injection into an analyser.

Simplifying UHPLC/HPLC sample preparation minimises instrument downtime resulting from use of unfiltered samples. Excessive leachables contamination from plastic vials can destabilise organic formulations and skew results. Mini-UniPrep™ G2 from Cytiva's business features a borosilicate glass chamber, so sample contact with plastic leachables is minimised.

With hand compressor.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge