Millex® Syringe Filters, PTFE Membrane
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SLFG65010 SLFG55010 SLFG05000 SLFG02550 SLFGL25BS SLFGN25VS SLFG025LS SLFA05000 SLFA05010 SLFG0250M SLFG65000 SLFG75000 SLFG75010 SLFH05000 SLFH05010 SLFG05010
MILFSLFG65010EA 185 EUR InStock
MILFSLFG65010 516-6551 MILFSLFG05000 514-0409 MILFSLFGL25BS MILFSLFGN25VS 514-0410 MILFSLFA05000 MILFSLFA05010 MILFSLFG0250M MILFSLFG65000 MILFSLFG75000 MILFSLFG75010 MILFSLFH05000 MILFSLFH05010 MILFSLFG05010
Millex® Syringe Filters, PTFE Membrane
Filter Spritzenvorsatzfilter
Syringe filters with hydrophobic PTFE membrane. Used in vacuum line protection, transducer protection and gas filtration. Available as either 25 mm or pump-driven Millex®-FG₅₀ 50 mm diameter filters.
Filters are either non sterile or sterilised with ethylene oxide.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge