Pipette controller, Falcon®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
357469
514-1457EA 370.59 EUR InStock
514-1457
Pipette controller, Falcon®
Pipetten Pipettierhelfer
This light, ergonomic, motorised pipette controller is designed for use with glass and plastic serological pipettes from 0,5 to 100 ml.
  • Ergonomic design for exceptional comfort
  • Nosepiece with autoclavable pipette holder and filter
  • Ergonomically positioned finger triggers provide comfortable grip
  • Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and viscosities
  • LCD display for continuous visualisation of speed, modes, and battery status
  • Easy access to the battery compartment
  • Filters, pipette holder and nosepiece are autoclavable
Lieferumfang: Supplied with a two-position charging stand, a universal battery charger, three batteries, two 0,2 μm and two 0,45 μm hydrophobic polytetrafluoroethylene (PTFE) filters.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge