Filterhalter SolVac®
Lieferant: Pall Laboratory
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4020
514-4001EA 516 EUR InStock
514-4001
Filterhalter SolVac®
Filterhalter
Simplifies clean-up and degassing of mobile phase solvents and other solutions. The versatile design fits most HPLC bottles, flasks, and containers, and eliminates the added steps of washing flasks and transferring mobile phase solvent from flask to reservoir.
  • Draws directly from HPLC solvent bottle. Less likely to spill aggressive solvents than glass funnels or disposable cups
  • Patented magnetic seal is reliable and leak proof. Eliminates the possibility of membrane shifting or tearing which can occur with aluminium clamps or threaded holders
Reusable, chemically resistant PP construction is resistant to common HPLC mobile phase solvents such as methanol, acetonitrile, and tetrahydrofuran.
Applications: Remove contaminating particulate from mobile phase or other solutions, de-gas mobile phase solvents and solutions, eliminate pour-and-wait filtration, remove microbial growth every 24 hours from aqueous buffer mobile phases.
Lieferumfang: Lieferung mit Ansaugschlauch (610 mm), Klemme, Sinker, Vakuumanschlussadapter, zwei Membran-Dichtungsmanschetten und zwei Dichtungen.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge