Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Wägepapiere, Sorte 2122, Whatman™

Lieferant: GE Healthcare - Whatman

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

10347890 10347893
515-8214EA 147 EUR InStock
515-8214 515-8212
Wägepapiere, Sorte 2122, Whatman™
Wägepapier
These Grade 2122 weigh paper sheets reliably allow samples to be transferred to scales without adding unwanted substances that could impact analytical results.

  • Smooth surface allows samples to slide off with ease and not be retained on paper
  • Shiny pergamyne paper allows visual determination that all sample has been removed
  • Extremely thin (0,03 mm) and lightweight (40 g/m²) for safe and secure sample transport
  • No glue or additives (residue-free)

This paper is not low in nitrogen. Therefore, it should not be used for Kjeldahl analysis.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge