Quarzmikrofaserfilter für die Luftprobenahme, Sorte QM-A, Whatman™
Lieferant: GE Healthcare - Whatman
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1851-025 1851-037 1851-047 1851-050 1851-055 1851-085 1851-865 1851-150 1851-032 1851-082 1851-090 1851-101 1851-110 1851-118 1851-8866 1851-045 1851-037_L
512-0430EA 240 EUR InStock
512-0430 516-2608 516-2611 516-5116 516-0199 516-5119 512-0431 516-5122 513-0112 512-0671 513-0086 513-0113 516-5120 516-5121 516-5123 WHAT1851-045 WHAT1851-037_L
Quarzmikrofaserfilter für die Luftprobenahme, Sorte QM-A, Whatman™
Filter Glasfaserfilter
These high purity quartz microfibre filters are used in many air sampling filter applications, including sampling of acidic gases, stacks, flues and aerosols. They are suitable for PM-10 sampling and analysis.
  • Binder-free
  • High loading capacity
Because of the low level of alkaline earth metals, 'artifact' products of sulfates and nitrates (from SO₂ and NO₂) are virtually eliminated. High-purity quartz microfiber filters are therefore applicable for particulate monitoring and heavy metal analysis.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge