Membrane filters, non sterile, ReliaDisc™
Lieferant: Ahlstrom-Munksjö
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
760211 760210 760102 760104 760105 760107 760150 760152 760153 760171 760181 760202 760203 760204 760206 760208 760209 760212 760106 760151 760207
516-0797EA 72.4 EUR InStock
516-0797 514-0732 516-0782 516-0783 516-0784 516-0785 516-0786 516-0787 516-0788 516-0789 516-0790 516-0791 516-0792 516-0793 516-0794 516-0795 516-0796 516-0798 514-0792 514-0793 514-0794
Membrane filters, non sterile, ReliaDisc™
Filter Membranfilter
These plain white non sterile microfiltration membranes are used to remove particles and microorganisms from various organic solvents and aqueous solutions.
  • Particle retention and collection
  • Sterile filtration of solutions
  • Particulate analysis
  • Air monitoring applications
  • Microbiological analysis of potable and waste waters
Cellulose acetate: Low protein-binding, well suited for sterile filtration and clarification of aqueous solutions in the pH range 4 - 8, nutrient media, buffers, and sera. Also compatible with alcohol and oil. These hydrophilic membranes offer high and consistent flow rates. Maximum thermal stability is 180 °C.
Cellulose nitrate: Set standard in both analytical and microbiological filtration. Available in a wide range of pore size and is well suited for microbial analysis in water, food and beverage application in the pH range 4 - 8. Maximum thermal stability is 130 °C. These membranes are available in a 47 and 50 mm ready-to-use standard size and it supports optimum colony growth.
Regenerated cellulose: Hydrophilic, pH range 3 to 12 and resistant to solvents. Typically used for HPLC sample preparation. Maximum thermal stability is 200 °C.
Nylon (Polyamide): Hydrophilic and widely used in both aqueous in the pH range 3 to 14 and organic solvent filtration applications. Well suited for clarification of buffers and nutrient media with a low level of extractables. Maximum thermal stability is 100 °C.
PTFE: Permanently hydrophobic filter material, very suitable for air and gas particulate filtration. Extremely resistant to aggressive solvents and acids in the pH range 1 to 14. Maximum thermal stability is 200 °C. Must be pre-wetted with ethanol before the filtration of aqueous media.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge