Rotors for Kr4i/Kr25i Centrifuges
Lieferant: JOUAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11177397 11177398 11178073 11178111 11178119 11178252 11178255 11177394 11177395 11177392 11178250 11178254 11178119
521-0611EA 7360 EUR InStock
521-0611 521-0612 JOUA11178073 521-0383 521-0384 521-0390 JOUA11178255 521-0608 521-0609 521-0606 JOUA11178250 521-0391 HERA11178119
Rotors for Kr4i/Kr25i Centrifuges
Zentrifugen Rotoren
Utilize the rotating capabilities of the Thermo Scientific™ RP6 Windshielded Swinging-Bucket Rotor, a high-capacity, low-speed rotor.
Capacity: 6×1000 ml (6 L) with Round bucket, 12×250 ml (3 L) with Rectangular bucket, Max. speed/RCF: 4250 rpm/5,735×g with Round bucket, 5,392×g with Rectangular rotor.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge