Rotor for Centrifuges 5418 R
Lieferant: EPPENDORF
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5418707.005
521-1519EA 627 EUR InStock
521-1519
Rotor for Centrifuges 5418 R
Zentrifugen Rotoren
Eppendorf rotor for centrifuges 5418 R, precision redefined. Tailor your centrifugation with varied capacities and speeds. Accessorise with adaptors and buckets for versatility in molecular biology and life science applications.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotor, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
Experience precision in centrifugation with Eppendorf rotor for centrifuges 5418 R. Offering a spectrum of capacities and speeds, these rotors cater to diverse applications from microcentrifugation to larger sample volumes. Accessorise your setup with adaptors and buckets, enhancing versatility for specific molecular biology and life science workflows. Achieve optimal sample separation, ensuring reproducibility in your experiments. Explore the array of options to customise your centrifuge 5418 R for reliable and efficient performance.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge