H-FLEX HS4 windshield swinging bucket rotor and accessories
Lieferant: HERAEUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
75003330 75003338 75003339
521-3440EA 4100 EUR InStock
521-3440 521-3441 521-3442
H-FLEX HS4 windshield swinging bucket rotor and accessories
Zentrifugen Standzentrifugen
Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor at 7197xg in 4 L High Performance General Purpose Centrifuges.
  • SMARTSpin™ technology
  • Space saving design
Swing bucket rotor enables fast Auto-Lock, one-handed rotor exchanges. Spins blood tubes, conical tubes and microplates without changing rotor or buckets. Results are faster without the need to change hardware. Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants.
Swing bucket rotor can be used with equipments such as Sorvall X4 Pro, Sorvall X4 F Pro, Multifuge X4 Pro, Multifuge X4 F Pro, Sorvall ST4 Plus, Sorvall ST4F Plus, Megafuge ST4 Plus, Megafuge ST4F Plus.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge