Fixed-angle rotor package, TLA-110
Lieferant: BECKMAN COULTER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
366735
521-4035EA 15500 EUR InStock
521-4035
Fixed-angle rotor package, TLA-110
Zentrifugen Rotoren
Major applications: Moderate volume differential sedimentation (pelleting) of subcellular organelles and viruses.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge