Potato native acid phosphatase
Lieferant: Calbiochem (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
524529-1
524529-1EA 579 EUR InStock
524529-1
Potato native acid phosphatase
Enzyme
Native potato acid phosphatase. Ideal for the removal of phosphate groups from phosphoproteins, such as β-casein, ovalbumin and pepsinogen.
One unit is defined as the amount of enzyme that will hydrolyse 1 µmol of pNPP/min at 25 °C, pH 5,6.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge