Buffer, PBS tablets, pH 7,4, Millipore®
Lieferant: Novabiochem (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
524650-1
524650-1EA 66.6 EUR InStock
524650-1
Buffer, PBS tablets, pH 7,4, Millipore®
Puffer Elektrophorese-Puffer Waschpuffer
Developed for use in immunoassay applications.
Dissolving one tablet of PBS buffer in 1 litre of deionised water yields 140 mM NaCl, 10 mM phosphate buffer, and 3 mM KCl, pH 7,4 at 25 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge