VWR®, Zentrifugenröhrchen
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
525-0638EA 88.6 EUR InStock
525-0638 525-0639 525-0640
VWR®, Zentrifugenröhrchen
Röhrchen Zentrifugenröhrchen
PS, glasklar, konisch, mit flachem Schraubverschluss mit integrierter Dichtung.
  • Electron beam irradiated
  • Longer length screw caps with sealing ring prevent any leakage
  • Engraved graduations at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge