Centrifuge tube, round bottom, with screw cap, KIMAX®, KIMBLE®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
45212-50 SP45212-50 SP45212-35
KIML45212-50EA 0 EUR InStock
KIML45212-50 525-0788 525-0787
Centrifuge tube, round bottom, with screw cap, KIMAX®, KIMBLE®
Röhrchen Zentrifugenröhrchen
Tube has a heavy wall to withstand higher speeds in centrifuging.
It is especially useful in separating neutralised sediment for culturing from sputum specimens, as well as many other clinical and microbiological applications.
Lieferumfang: Supplied with unattached autoclavable cap with cemented-in rubber liner.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge